Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(5-Methyl-1,2,4-oxadiazol-3-yl)aniline, 97%, Thermo Scientific™

Catalog No. CC39114DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC39114DE 5 g
CC39114CB 250 mg
CC39114DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC39114DE Supplier Thermo Scientific Chemicals Supplier No. CC39114DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10185-69-0
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.19
MDL Number MFCD08443707
InChI Key CTRGRIHPFAVSOF-UHFFFAOYSA-N
Synonym 3-5-methyl-1,2,4-oxadiazol-3-yl aniline, 3-5-methyl-1,2,4-oxadiazol-3-yl phenylamine, benzenamine,3-5-methyl-1,2,4-oxadiazol-3-yl
PubChem CID 11423869
IUPAC Name 3-(5-methyl-1,2,4-oxadiazol-3-yl)aniline
SMILES CC1=NC(=NO1)C1=CC(N)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 3-(5-Methyl-1, 2, 4-oxadiazol-3-yl)aniline
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.