CAS RN 1019-57-4
IUPAC Name:
(2E)-1-(2,4-dinitrophenyl)-2-ethylidenehydrazine
Synonyms:
(2E)-1-(2,4-dinitrophenyl)-2-ethylidenehydrazine
Molecular Weight (g/mol):
224.18
Molecular Formula:
C8H8N4O4
InChi Key:
ONBOQRNOMHHDFB-XNWCZRBMSA-N
SMILES:
C\C=N\NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O