Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Acetaldehyde-2,4-DNPH Solution, 1,000 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111018976 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8976 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8976 Supplier MilliporeSigma Supelco Supplier No. CRM4M7340
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Packaging Ampule
CAS 1019-57-4
Product Line TraceCERT
Flash Point 2.0°C - closed cup
Concentration 1000 μg/mL in acetonitrile (as the aldehyde)
Linear Formula C8H8N4O4
Synonym Acetaldehyde-2,4-dinitrophenylhydrazone solution
Type Organic Pollutant Standard
Quantity 5 x 1 mL
Formula Weight 224.17
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.