accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaCH3OHNONNSOOOSCH3SOOONCH3O(E/Z)

CAS RN 10190-68-8

IUPAC Name: disodium 2-[4-(2-{1-[(2-methoxyphenyl)carbamoyl]-2-oxopropyl}diazen-1-yl)-3-sulfonatophenyl]-6-methyl-1,3-benzothiazole-7-sulfonate
Synonyms: disodium 2-[4-(2-{1-[(2-methoxyphenyl)carbamoyl]-2-oxopropyl}diazen-1-yl)-3-sulfonatophenyl]-6-methyl-1,3-benzothiazole-7-sulfonate
Molecular Weight (g/mol): 662.61
Molecular Formula: C25H20N4Na2O9S3
InChi Key: WLDNGJFRVWQASY-UHFFFAOYNA-L
SMILES: [Na+].[Na+].COC1=CC=CC=C1NC(=O)C(N=NC1=CC=C(C=C1S([O-])(=O)=O)C1=NC2=CC=C(C)C(=C2S1)S([O-])(=O)=O)C(C)=O
Color 
  • (1)
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)