Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Direct Yellow 27, pure, Thermo Scientific™

Catalog No. AC191840500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC191840500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC191840500 Supplier Thermo Scientific Chemicals Supplier No. 191840500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10190-68-8
Molecular Formula C25H22N4Na2O9S3
Molecular Weight (g/mol) 664.63
InChI Key GTJLVZCXBWMOAC-UHFFFAOYSA-N
Synonym C.I. 13950
PubChem CID 131843060
IUPAC Name 2-[4-[[1-(2-methoxyanilino)-1,3-dioxobutan-2-yl]diazenyl]-3-sulfophenyl]-6-methyl-1,3-benzothiazole-7-sulfonic acid;sodium
SMILES CC1=C(C2=C(C=C1)N=C(S2)C3=CC(=C(C=C3)N=NC(C(=O)C)C(=O)NC4=CC=CC=C4OC)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]

Specifications

Infrared Spectrum Authentic
Lambda Max 392 to 394nm
Formula Weight 662.6
Grade Extra Pure
Color Orange to Yellow
Quantity 50 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Direct yellow 27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.