accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1019205-65-2

IUPAC Name: bromozincylium; cyclobutanide
Synonyms: bromozincylium; cyclobutanide
Molecular Weight (g/mol): 200.38
Molecular Formula: C4H7BrZn
InChi Key: WNFQEBSVYUDIES-UHFFFAOYSA-M
SMILES: [Zn+]Br.C1C[CH-]C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results