Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclobutylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58247AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58247AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58247AD Supplier Thermo Scientific Chemicals Supplier No. H58247AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1019205-65-2 | C4H7BrZn | 200.384 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This compound can be employed for cross coupling with acid chlorides, aryl halides, and substituted or non-substituted allyl halides. They may also undergo Michael Addition to unsaturated ketones or additions to alkynes to give subs
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1019205-65-2
Molecular Formula C4H7BrZn
Molecular Weight (g/mol) 200.384
MDL Number MFCD09801445
InChI Key WNFQEBSVYUDIES-UHFFFAOYSA-M
Synonym cyclobutylzinc bromide, cyclobutylzinc bromide, 0.5 m in thf, cyclobutylzinc bromide 0.5m in tetrahydrofuran, cyclobutylzinc bromide, 0.5m in tetrahydrofuran
PubChem CID 24722675
IUPAC Name bromozinc(1+);cyclobutane
SMILES C1C[CH-]C1.[Zn+]Br

Specifications

Density 0.984
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Solubility Information Reacts with water.
Formula Weight 200.38
Chemical Name or Material Cyclobutylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H260-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.