CAS RN 1019642-47-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ CD22L-N3 (BPC-Sia-LacNAc-N3)
CD22 ligand with azide group for onward chemistry