accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102-02-3

IUPAC Name: hydrogen N-(diaminomethylidene)-N''-phenylguanidine chloride
Synonyms: phenylbiguanide
Molecular Weight (g/mol): 213.67
Molecular Formula: C8H12ClN5
InChi Key: FHUDRDSKZQDCBC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(N)=NC(N)=NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results