accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3C

CAS RN 102-25-0

IUPAC Name: 1,3,5-triethylbenzene
Synonyms: 1,3,5-triethylbenzene
Molecular Weight (g/mol): 162.28
Molecular Formula: C12H18
InChi Key: WJYMPXJVHNDZHD-UHFFFAOYSA-N
SMILES: CCC1=CC(CC)=CC(CC)=C1
Boiling Point 
  • (3)
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (7)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)