Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Triethylbenzene 90.0+%, TCI America™
SDP

Catalog No. T2352500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2352500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2352500ML Supplier TCI America Supplier No. T2352500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 102-25-0
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00009261
InChI Key WJYMPXJVHNDZHD-UHFFFAOYSA-N
Synonym benzene, 1,3,5-triethyl, unii-02k328wwzq, 1,5-triethylbenzene, pubchem19794, benzene,3,5-triethyl, 1,3,5-triethyl benzene, ksc493q9l, wjympxjvhndzhd-uhfffaoysa, 1,3,5-triethylbenzene, 1,3,5-triethylbenzene, technical grade
PubChem CID 7602
IUPAC Name 1,3,5-triethylbenzene
SMILES CCC1=CC(CC)=CC(CC)=C1

Specifications

Melting Point -66°C
Color Colorless
Boiling Point 215°C
Quantity 500 mL
UN Number 3082
Formula Weight 162.28
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Triethylbenzene
EINECSNumber (3)-3427
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.