accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102-50-1

IUPAC Name: 4-methoxy-2-methylaniline
Synonyms: 4-methoxy-2-methylaniline
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: CDGNLUSBENXDGG-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C(C)=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)