Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-2-methylaniline, 98%, Thermo Scientific™

Catalog No. AAL1491022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1491022 100 g
AAL1491006 5 g
AAL1491014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1491022 Supplier Thermo Scientific Chemicals Supplier No. L1491022

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-50-1
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00007735
InChI Key CDGNLUSBENXDGG-UHFFFAOYSA-N
Synonym m-cresidine, 2-methyl-4-methoxyaniline, 2-methyl-p-anisidine, 4-methoxy-o-toluidine, benzenamine, 4-methoxy-2-methyl, meta-cresidine, p-anisidine, 2-methyl, 6-amino-3-methoxytoluene, 2-methyl-4-methoxybenzenamine, 4-methoxy-2-methylbenzenamine
PubChem CID 7610
ChEBI CHEBI:82430
IUPAC Name 4-methoxy-2-methylaniline
SMILES CC1=C(C=CC(=C1)OC)N

Specifications

Melting Point 13°C to 14°C
Density 1.065
Boiling Point 248°C to 249°C
Flash Point 124°C (255°F)
Refractive Index 1.5685
Quantity 100 g
UN Number UN2810
Beilstein 774727
Formula Weight 137.18
Percent Purity 98%
Chemical Name or Material 4-Methoxy-2-methylaniline
Show More Show Less
Hazard Category H315-H319-H335-H351
Hazard Statement GHS H Statement
H331-H351-H302-H312
Toxic if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 203-036-7
RTECSNumber BZ6730000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.