accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102-60-3

IUPAC Name: 1-({2-[bis(2-hydroxypropyl)amino]ethyl}(2-hydroxypropyl)amino)propan-2-ol
Synonyms: quadrol tetrahydroxypropyl ethylenediamine entprol neutrol TE edetolum quadrol L edetol adeka quadrol THPE
Molecular Weight (g/mol): 292.42
Molecular Formula: C14H32N2O4
InChi Key: NSOXQYCFHDMMGV-UHFFFAOYNA-N
SMILES: CC(O)CN(CCN(CC(C)O)CC(C)O)CC(C)O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (1)
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (4)