Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine 98%, Thermo Scientific™

Catalog No. AC153422500
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153422500 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC153422500 Supplier Thermo Scientific Chemicals Supplier No. 153422500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-60-3
Molecular Formula C14H32N2O4
Molecular Weight (g/mol) 292.42
MDL Number MFCD00004534
InChI Key NSOXQYCFHDMMGV-UHFFFAOYSA-N
Synonym quadrol, edetol, n,n,n',n'-tetrakis 2-hydroxypropyl ethylenediamine, entprol, adeka quadrol, quadrol l, neutrol te, 1,1',1,1'-ethylenedinitrilo tetra-2-propanol, edetolum latin, 2-propanol, 1,1',1,1'-1,2-ethanediyldinitrilo tetrakis
PubChem CID 7615
IUPAC Name 1-[2-[bis(2-hydroxypropyl)amino]ethyl-(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(CN(CCN(CC(C)O)CC(C)O)CC(C)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.01g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Refractive Index 1.4795 to 1.4815 (20°C, 589nm)
Linear Formula [CH3CH(OH)CH2]2NCH2CH2N[CH2CH(OH)CH3]2
Quantity 250 mL
Beilstein 04, IV, 1685
Merck Index 15, 3653
Formula Weight 292.41
Percent Purity ≥97.5%
Physical Form Viscous Liquid
Chemical Name or Material N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine, 98%
Show More Show Less
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.