accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102-87-4

IUPAC Name: tridodecylamine
Synonyms: trilaurylamine
Molecular Weight (g/mol): 522
Molecular Formula: C36H75N
InChi Key: SWZDQOUHBYYPJD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCN(CCCCCCCCCCCC)CCCCCCCCCCCC
Boiling Point 
  • (1)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)