Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tridodecylamine, TCI America™
SDP

Catalog No. T0634500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0634500ML 500 mL
T063425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0634500ML Supplier TCI America Supplier No. T0634500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 102-87-4
Molecular Formula C36H75N
Molecular Weight (g/mol) 522.003
MDL Number MFCD00008971
InChI Key SWZDQOUHBYYPJD-UHFFFAOYSA-N
Synonym tridodecylamine, trilaurylamine, tri-n-dodecylamine, alamine 304, tridodecyl amine, adogen 360, 1-dodecanamine, n,n-didodecyl, n,n-didodceyl-1-dodecanamine, n,n-didodecyl-1-dodecanamine, unii-sf36t21u17
PubChem CID 7624
IUPAC Name N,N-didodecyldodecan-1-amine
SMILES CCCCCCCCCCCCN(CCCCCCCCCCCC)CCCCCCCCCCCC

Specifications

Melting Point 16°C
Color Yellow
Boiling Point 296°C
Quantity 500 mL
Formula Weight 522.00
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tridodecylamine
EINECSNumber (2)-0176
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.