accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBrNOOOOH

CAS RN 1020717-99-0

IUPAC Name: 4-bromo-5-fluoro-2-nitrobenzoic acid
Synonyms: 4-bromo-5-fluoro-2-nitrobenzoic acid
Molecular Weight (g/mol): 264.01
Molecular Formula: C7H3BrFNO4
InChi Key: JZVCMOJSPCGLTC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=C(Br)C=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results