CAS RN 1021176-69-1
CAS RN 1021176-69-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich (S)-(+)-N-(3,5-Dioxa-4-Phosphacyclohepta[2,1-A;3,4-A']Dinaphthalen-4-Yl)-Dibenzo[B,F]Azepine
Sigma Aldrich (S)-(+)-N-(3,5-Dioxa-4-Phosphacyclohepta[2,1-A;3,4-A']Dinaphthalen-4-Yl)-Dibenzo[B,F]Azepine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.