CAS RN 10213-78-2
IUPAC Name:
2-[(2-hydroxyethyl)(octadecyl)amino]ethan-1-ol
Synonyms:
peg-2 stearamine
Molecular Weight (g/mol):
357.62
Molecular Formula:
C22H47NO2
InChi Key:
NDLNTMNRNCENRZ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCN(CCO)CCO
1
–
1
of
1
results
1
Stearyldiethanolamine 98.0+%, TCI America™
CAS: 10213-78-2 Molecular Formula: C22H47NO2 Molecular Weight (g/mol): 357.623 MDL Number: MFCD00059003 InChI Key: NDLNTMNRNCENRZ-UHFFFAOYSA-N PubChem CID: 61494 IUPAC Name: 2-[2-hydroxyethyl(octadecyl)amino]ethanol SMILES: CCCCCCCCCCCCCCCCCCN(CCO)CCO