Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearyldiethanolamine 98.0+%, TCI America™
SDP

Catalog No. O00175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00175G 5 g
O001725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O00175G Supplier TCI America Supplier No. O00175G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10213-78-2
Molecular Formula C22H47NO2
Molecular Weight (g/mol) 357.623
MDL Number MFCD00059003
InChI Key NDLNTMNRNCENRZ-UHFFFAOYSA-N
PubChem CID 61494
IUPAC Name 2-[2-hydroxyethyl(octadecyl)amino]ethanol
SMILES CCCCCCCCCCCCCCCCCCN(CCO)CCO

Specifications

Melting Point 51°C
Color White
Quantity 5 g
Synonym 2,2′C-(Octadecylimino)diethanol
Formula Weight 357.62
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Stearyldiethanolamine
EINECSNumber (2)-0353&(7)-0060
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.