accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10214-40-1

CuOSeOO

CAS RN 10214-40-1

IUPAC Name: copper(2+) selenite
Synonyms: copper(II) selenite
Molecular Weight (g/mol): 190.51
Molecular Formula: CuO3Se
InChi Key: JNRBLFVQPTZFAG-UHFFFAOYSA-L
SMILES: [Cu++].[O-][Se]([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results