Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) selenite hydrate, 97%, Thermo Scientific™

Catalog No. AA4022130
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4022130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4022130 Supplier Thermo Scientific Chemicals Supplier No. 4022130

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10214-40-1
Molecular Formula CuO3Se
Molecular Weight (g/mol) 190.51
MDL Number MFCD00010973
InChI Key JNRBLFVQPTZFAG-UHFFFAOYSA-L
IUPAC Name copper(2+) selenite
SMILES [Cu++].[O-][Se]([O-])=O

Specifications

Color Blue
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 97%
Solubility Information Insoluble in water. Soluble in acids,NH4OH
Formula Weight 190.50 (Anhydrous)
Appearance Blue crystalline powder
Chemical Name or Material Copper(II) selenite hydrate
Hazard Category H301+H331-H373
Precautionary Statement P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
DOTInformation DOT Class: 6.1, Packing Group: I
EINECSNumber 233-526-6
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.