accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONHFBr

CAS RN 1021439-52-0

IUPAC Name: N-(2-bromo-4-fluorophenyl)-2-methoxybenzamide
Synonyms: N-(2-bromo-4-fluorophenyl)-2-methoxybenzamide
Molecular Weight (g/mol): 324.15
Molecular Formula: C14H11BrFNO2
InChi Key: CVEPLYNHBLSXCV-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)NC1=C(Br)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results