accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNHNSOOONOOSOOOOOOPOOONaNaNaNa(E/Z)

CAS RN 1021868-77-8

IUPAC Name: tetrasodium 3-(2-{4-formyl-6-methyl-5-oxo-3-[(phosphonatooxy)methyl]-2,5-dihydropyridin-2-ylidene}hydrazin-1-yl)-7-nitronaphthalene-1,5-disulfonate
Synonyms: tetrasodium 3-(2-{4-formyl-6-methyl-5-oxo-3-[(phosphonatooxy)methyl]pyridin-2-ylidene}hydrazin-1-yl)-7-nitronaphthalene-1,5-disulfonate
Molecular Weight (g/mol): 694.35
Molecular Formula: C18H11N4Na4O14PS2
InChi Key: TXFLDKHOZFWVAC-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].CC1=NC(=NNC2=CC(=C3C=C(C=C(C3=C2)S([O-])(=O)=O)[N+]([O-])=O)S([O-])(=O)=O)C(COP([O-])([O-])=O)=C(C=O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results