CAS RN 1021926-22-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Nimesulide Analog, Compound 76
A cell-permeable sulfonanilide that is shown to exhibit an antiproliferative effect against SKBR-3, BT474 and MDA-MB-453, and affects MCF-7 and MDA-MB-231 only at much higher concentrations.