Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Nimesulide Analog, Compound 76

Catalog No. 4822105MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-221-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-221-05MG Supplier MilliporeSigma™ Supplier No. 4822105MG
Add to Cart
Add to Cart

A cell-permeable sulfonanilide that is shown to exhibit an antiproliferative effect against SKBR-3, BT474 and MDA-MB-453, and affects MCF-7 and MDA-MB-231 only at much higher concentrations.

A cell-permeable sulfonanilide that is shown to exhibit an antiproliferative effect against three HER-2-positive breast cancer cell lines, SKBR-3, BT474 and MDA-MB-453 (IC50= 0.9, 2.2, and 4.0μM, respectively), and affects two HER-2-negative breast cancer cell lines, MCF-7 and MDA-MB-231 only at much higher concentrations (IC50 = 22.1 and 19.6μM, respectively). The compound is not expected to be a COX-2 inhibitor and its cellular target(s) and mechanism(s) of growth inhibition is yet to be seen.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
CAS 1021926-22-6
Molecular Formula C24H32N2O4S
Formula Weight 444.59g/mol
Solubility DMSO
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.