accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOHO

CAS RN 10223-33-3

IUPAC Name: 4-(2-hydroxyethyl)-3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 4-(2-hydroxyethyl)-5-methyl-2-phenyl-4H-pyrazol-3-one
Molecular Weight (g/mol): 218.26
Molecular Formula: C12H14N2O2
InChi Key: AVOONTISHAAFGN-UHFFFAOYNA-N
SMILES: CC1=NN(C(=O)C1CCO)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results