accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10241-05-1

IUPAC Name: molybdenum(5+) pentachloride
Synonyms: molybdenum(5+) pentachloride
Molecular Weight (g/mol): 273.2
Molecular Formula: Cl5Mo
InChi Key: GICWIDZXWJGTCI-UHFFFAOYSA-I
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5]
Molecular Weight (g/mol) 
  • (8)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (4)
  • (2)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (7)
  • (8)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (4)
  • (3)