Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum Pentachloride, MP Biomedicals

Catalog No. ICN210551 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210551 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210551 Supplier MP Biomedicals, Inc Supplier No. 0521055180

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10241-05-1
Molecular Formula Cl5Mo
Molecular Weight (g/mol) 273.20
MDL Number MFCD00003467
InChI Key GICWIDZXWJGTCI-UHFFFAOYSA-I
Synonym molybdenum v chloride, molybdenum pentachloride, molybdenum chloride mocl5, mocl5, pentachloridomolybdenum, molybdenum 5+ chloride, acmc-1bx8l, ksc491a7h
PubChem CID 61497
ChEBI CHEBI:30635
IUPAC Name molybdenum(5+) pentachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5]

Specifications

Quantity 100 g
Formula Weight 273.21
Chemical Name or Material Molybdenum Pentachloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.