accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1025113-78-3

BSOFFFK

CAS RN 1025113-78-3

IUPAC Name: potassium trifluoro(5-formylthiophen-2-yl)boranuide
Synonyms: potassium trifluoro(5-formylthiophen-2-yl)boranuide
Molecular Weight (g/mol): 218.04
Molecular Formula: C5H3BF3KOS
InChi Key: XERVPXDMNWPUFB-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=C(S1)C=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results