Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium 5-formylthiophene-2-trifluoroborate, Thermo Scientific™

Catalog No. AAH5114006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5114006 5 g
AAH5114003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5114006 Supplier Thermo Scientific Chemicals Supplier No. H5114006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1025113-78-3
Molecular Formula C5H3BF3KOS
Molecular Weight (g/mol) 218.042
MDL Number MFCD09800740
InChI Key XERVPXDMNWPUFB-UHFFFAOYSA-N
Synonym potassium 5-formylthiophene-2-trifluoroborate, potassium 5-formyl-2-thiophenetrifluoroborate, potassium trifluoro 5-formylthiophen-2-yl boranuide, amtb086, potassium trifluoro 5-formylthiophen-2-yl borate, potassium trifluoro 5-formylthiophen-2-yl borate 1-
PubChem CID 45479878
IUPAC Name potassium;trifluoro-(5-formylthiophen-2-yl)boranuide
SMILES [B-](C1=CC=C(S1)C=O)(F)(F)F.[K+]

Specifications

Quantity 5 g
Formula Weight 218.05
Chemical Name or Material Potassium 5-formylthiophene-2-trifluoroborate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.