accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CBrOOCH3

CAS RN 1025718-78-8

IUPAC Name: methyl 3-bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Synonyms: methyl 3-bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Molecular Weight (g/mol): 341.01
Molecular Formula: C14H18BBrO4
InChi Key: YTLNMWYKZKGCOV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results