Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-(methoxycarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog No. AAH51718MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51718MD 250 mg
AAH5171803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51718MD Supplier Thermo Scientific Chemicals Supplier No. H51718MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1025718-78-8
Molecular Formula C14H18BBrO4
Molecular Weight (g/mol) 341.008
MDL Number MFCD12405334
InChI Key YTLNMWYKZKGCOV-UHFFFAOYSA-N
Synonym methyl 3-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 3-bromo-5-methoxycarbonyl benzeneboronic acid pinacol ester, 3-bromo-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzoic acid methyl ester, 3-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzoic acid methyl ester, 3-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzoic acid, methyl ester, benzoic acid, 3-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, methyl ester
PubChem CID 53436914
IUPAC Name methyl 3-bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)Br)C(=O)OC

Specifications

Melting Point 98°C to 102°C
Quantity 250 mg
Formula Weight 341.01
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(methoxycarbonyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.