CAS RN 102649-78-5
IUPAC Name:
5-chloro-N-(6-phenylhexyl)naphthalene-1-sulfonamide
Synonyms:
5-chloro-N-(6-phenylhexyl)naphthalene-1-sulfonamide
Molecular Weight (g/mol):
401.95
Molecular Formula:
C22H24ClNO2S
InChi Key:
YUXMTDYHFPRPLG-UHFFFAOYSA-N
SMILES:
ClC1=C2C=CC=C(C2=CC=C1)S(=O)(=O)NCCCCCCC1=CC=CC=C1
1
–
1
of
1
results
1
SC-9, Tocris Bioscience™
CAS: 102649-78-5 Molecular Formula: C22H24ClNO2S Molecular Weight (g/mol): 401.95 MDL Number: MFCD00065544 InChI Key: YUXMTDYHFPRPLG-UHFFFAOYSA-N Synonym: sc-9,5-chloro-n-6-phenylhexyl-1-naphthalenesulfonamide,5-chloro-n-6-phenylhexyl naphthalene-1-sulfonamide,n-6-phenylhexyl-5-chloro-1-naphthalenesulfonamide,sc9,6-phcns,acmc-20eftt,tocris-0433,n-6-phenylhexyl-5-chloro-1-naphthalene-sulphonamide,1-naphthalenesulfonamide, 5-chloro-n-6-phenylhexyl PubChem CID: 124172 IUPAC Name: 5-chloro-N-(6-phenylhexyl)naphthalene-1-sulfonamide SMILES: ClC1=C2C=CC=C(C2=CC=C1)S(=O)(=O)NCCCCCCC1=CC=CC=C1