accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2

CAS RN 102653-37-2

IUPAC Name: 4-propylcyclohexan-1-amine
Synonyms: 4-propylcyclohexan-1-amine
Molecular Weight (g/mol): 141.26
Molecular Formula: C9H19N
InChi Key: OZUBMBIDHPBIDL-UHFFFAOYSA-N
SMILES: CCCC1CCC(N)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results