Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propylcyclohexylamine (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. P20225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20225ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P20225ML Supplier TCI America Supplier No. P20225ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 102653-37-2
Molecular Formula C9H19N
Molecular Weight (g/mol) 141.258
MDL Number MFCD11156904
InChI Key OZUBMBIDHPBIDL-UHFFFAOYSA-N
Synonym 1-Amino-4-propylcyclohexane
PubChem CID 544651
IUPAC Name 4-propylcyclohexan-1-amine
SMILES CCCC1CCC(CC1)N

Specifications

Color Colorless
Quantity 5 mL
UN Number 2733
Formula Weight 141.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Propylcyclohexylamine (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.