accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNIOHIOHO

CAS RN 1027-28-7

IUPAC Name: 2-acetamido-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
Synonyms: 2-acetamido-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
Molecular Weight (g/mol): 475.02
Molecular Formula: C11H11I2NO4
InChi Key: CDXURJOCZAIXFK-UHFFFAOYNA-N
SMILES: CC(=O)NC(CC1=CC(I)=C(O)C(I)=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results