Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Acetyl-3,5-diiodo-L-tyrosine 95.0+%, TCI America™
SDP

Catalog No. A0087100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0087100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0087100MG Supplier TCI America Supplier No. A0087100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1027-28-7
Molecular Formula C11H11I2NO4
Molecular Weight (g/mol) 475.021
MDL Number MFCD00020004
InChI Key CDXURJOCZAIXFK-VIFPVBQESA-N
PubChem CID 7171490
IUPAC Name (2S)-2-acetamido-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
SMILES CC(=O)NC(CC1=CC(=C(C(=C1)I)O)I)C(=O)O

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 100 mg
Formula Weight 475.02
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-3,5-diiodo-L-tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.