CAS RN 102830-49-9
CAS RN 102830-49-9
IUPAC Name:
3-(benzylsulfanyl)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms:
3-(benzylsulfanyl)-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol):
311.4
Molecular Formula:
C15H21NO4S
InChi Key:
IFVORPLRHYROAA-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC(CSCC1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Boc-D-Cys Bzl -OH, Advanced ChemTech
CAS: 102830-49-9 Molecular Formula: C15H21NO4S Molecular Weight (g/mol): 311.40 InChI Key: IFVORPLRHYROAA-UHFFFAOYNA-N IUPAC Name: 3-(benzylsulfanyl)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid SMILES: CC(C)(C)OC(=O)NC(CSCC1=CC=CC=C1)C(O)=O