accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102936-05-0

IUPAC Name: (2S)-2-[(phenylcarbamoyl)oxy]propanoate
Synonyms: (2S)-2-[(phenylcarbamoyl)oxy]propanoate
Molecular Weight (g/mol): 208.19
Molecular Formula: C10H10NO4
InChi Key: HYIHYWQSOXTJFS-ZETCQYMHSA-M
SMILES: C[C@H](OC(=O)NC1=CC=CC=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results