Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-[(Phenylamino)carbonyloxy]propionic acid, 99%, Thermo Scientific™

Catalog No. AC235290100
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC235290100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC235290100 Supplier Thermo Scientific Chemicals Supplier No. 235290100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 102936-05-0
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C10H11NO4
Quantity 10g
MDL Number MFCD00063138
Specific Rotation Condition − 12.50 (20.00°C c=1,C2H5OH)
Specific Rotation -11° to -14° (20°C, 589nm) (c=1, C2H5OH)
Synonym s---2-phenylcarbamoyloxy propionic acid, s-2-phenylcarbamoyl oxy propanoic acid, s---2-phenylamino carbonyloxy propionic acid, 2s-2-phenylcarbamoyl oxy propanoic acid, 2s-2-n-phenylcarbamoyloxy propanoic acid, pubchem16496, s-pacopa, s-2-phenylcarbamoyloxy propionic acid, 2s-2-phenylcarbamoyloxy propanoic acid, s-2-phenylcarbamoyloxy propanoic acid
InChI Key HYIHYWQSOXTJFS-ZETCQYMHSA-N
SMILES CC(C(=O)O)OC(=O)NC1=CC=CC=C1
IUPAC Name (2S)-2-(phenylcarbamoyloxy)propanoic acid
Molecular Weight (g/mol) 209.201
PubChem CID 736218
Formula Weight 209.2
Percent Purity ≥98.5%
Chemical Name or Material (S)-(-)-2-[(Phenylamino)carbonyloxy]propionic acid, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.