CAS RN 10294-26-5
Silver Sulfate (Powder/Certified ACS), Fisher Chemical™
CAS: 10294-26-5 Molecular Formula: Ag2H2O4S Molecular Weight (g/mol): 313.81 MDL Number: MFCD00003407 InChI Key: YPNVIBVEFVRZPJ-UHFFFAOYSA-N Synonym: silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt PubChem CID: 159865 IUPAC Name: disilver(1+) sulfuric acid SMILES: [Ag+].[Ag+].OS(O)(=O)=O
Silver sulfate, Premion™, 99.999% (metals basis)
CAS: 10294-26-5 Molecular Formula: Ag2H2O4S Molecular Weight (g/mol): 313.81 MDL Number: MFCD00003407 InChI Key: YPNVIBVEFVRZPJ-UHFFFAOYSA-N Synonym: silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt PubChem CID: 159865 IUPAC Name: disilver(1+) sulfuric acid SMILES: [Ag+].[Ag+].OS(O)(=O)=O
Silver sulfate, ACS, 98% min
CAS: 10294-26-5 Molecular Formula: Ag2H2O4S Molecular Weight (g/mol): 313.81 MDL Number: MFCD00003407 InChI Key: YPNVIBVEFVRZPJ-UHFFFAOYSA-N Synonym: silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt PubChem CID: 159865 IUPAC Name: disilver(1+) sulfuric acid SMILES: [Ag+].[Ag+].OS(O)(=O)=O
Silver sulfate, pure, 10 g/l solution in sulfuric acid
CAS: 10294-26-5 | Ag2H2O4S | 313.81 g/mol
Silver sulfate, Premion™, 99.99% (metals basis), Ag 68.9% min
CAS: 10294-26-5 Molecular Formula: Ag2H2O4S Molecular Weight (g/mol): 313.81 MDL Number: MFCD00003407 InChI Key: YPNVIBVEFVRZPJ-UHFFFAOYSA-N Synonym: silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt PubChem CID: 159865 IUPAC Name: disilver(1+) sulfuric acid SMILES: [Ag+].[Ag+].OS(O)(=O)=O
Silver Sulfate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 10294-26-5 Molecular Formula: Ag2H2O4S Molecular Weight (g/mol): 313.81 InChI Key: YPNVIBVEFVRZPJ-UHFFFAOYSA-N Synonym: silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt PubChem CID: 159865 IUPAC Name: disilver(1+) sulfuric acid SMILES: [Ag+].[Ag+].OS(O)(=O)=O
Silver sulfate, ACS reagent
CAS: 10294-26-5 Molecular Formula: Ag2H2O4S Molecular Weight (g/mol): 313.81 InChI Key: YPNVIBVEFVRZPJ-UHFFFAOYSA-N Synonym: silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt PubChem CID: 159865 IUPAC Name: disilver(1+) sulfuric acid SMILES: [Ag+].[Ag+].OS(O)(=O)=O
Silver sulfate, 99%, extra pure
CAS: 10294-26-5 Molecular Formula: Ag2H2O4S Molecular Weight (g/mol): 313.81 MDL Number: MFCD00003407 InChI Key: YPNVIBVEFVRZPJ-UHFFFAOYSA-N Synonym: silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt PubChem CID: 159865 IUPAC Name: disilver(1+) sulfuric acid SMILES: [Ag+].[Ag+].OS(O)(=O)=O
Silver Sulfate, ACS Reagent Grade, Ricca Chemical
CAS: 10294-26-5
Silver Sulfate, ACS Grade, ≥ 98.0%, LabChem™
CAS: 10294-26-5 Molecular Formula: Ag2H2O4S Molecular Weight (g/mol): 313.81 InChI Key: YPNVIBVEFVRZPJ-UHFFFAOYSA-N Synonym: silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt PubChem CID: 159865 IUPAC Name: disilver(1+) sulfuric acid SMILES: [Ag+].[Ag+].OS(O)(=O)=O
Silver Sulfate-Sulfuric Acid, For COD, LabChem™
CAS: 10294-26-5 Molecular Formula: Ag2H2O4S Molecular Weight (g/mol): 313.81 InChI Key: YPNVIBVEFVRZPJ-UHFFFAOYSA-N Synonym: silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt PubChem CID: 159865 IUPAC Name: disilver(1+) sulfuric acid SMILES: [Ag+].[Ag+].OS(O)(=O)=O
Silver Sulfate, puriss. p.a. ACS Reagent, Solstice
CAS: 10294-26-5 Molecular Formula: Ag2H2O4S Molecular Weight (g/mol): 313.81 MDL Number: MFCD00003407 InChI Key: YPNVIBVEFVRZPJ-UHFFFAOYSA-N Synonym: silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt PubChem CID: 159865 IUPAC Name: disilver(1+) sulfuric acid SMILES: [Ag+].[Ag+].OS(O)(=O)=O
Silver Sulfate, puriss., Solstice
CAS: 10294-26-5 Molecular Formula: Ag2H2O4S Molecular Weight (g/mol): 313.81 MDL Number: MFCD00003407 InChI Key: YPNVIBVEFVRZPJ-UHFFFAOYSA-N Synonym: silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt PubChem CID: 159865 IUPAC Name: disilver(1+) sulfuric acid SMILES: [Ag+].[Ag+].OS(O)(=O)=O
Sigma Aldrich Silver Sulfate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Silver Sulfate, 98.0% min., ACS, MilliporeSigma™
CAS: 10294-26-5 Molecular Formula: Ag2H2O4S Molecular Weight (g/mol): 313.81 InChI Key: YPNVIBVEFVRZPJ-UHFFFAOYSA-N Synonym: silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt PubChem CID: 159865 IUPAC Name: disilver(1+) sulfuric acid SMILES: [Ag+].[Ag+].OS(O)(=O)=O
- 1
- 2