Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Sulfate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCS0200500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCS0200500 500 g
RDCS020025F 25 g
RDCS0200125 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RDCS0200500 Supplier Ricca Chemical Company Supplier No. RDCS0200500C2
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 10294-26-5

Specifications

CAS Max % 100
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Molecular Formula Ag2H2O4S
InChI Key YPNVIBVEFVRZPJ-UHFFFAOYSA-N
SMILES [Ag+].[Ag+].OS(O)(=O)=O
IUPAC Name disilver(1+) sulfuric acid
Molecular Weight (g/mol) 313.81
Percent Purity min 98%
Grade ACS
Packaging Amber Glass Bottle
Chemical Name or Material SIlver Sulfate
Show More Show Less
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.