accessibility menu, dialog, popup

UserName

Viewing profile as user:
KKOSOOS

CAS RN 10294-66-3

IUPAC Name: dipotassium 1-oxo-1λ⁶-disulfene-1,1-bis(olate)
Synonyms: potassium thiosulfate
Molecular Weight (g/mol): 190.31
Molecular Formula: K2O3S2
InChi Key: FGRVOLIFQGXPCT-UHFFFAOYSA-L
SMILES: [K+].[K+].[O-]S([O-])(=O)=S
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)