Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Potassium Thiosulfate

Catalog No. 12669500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-669-500G 500 g
12-669-100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-669-500G Supplier Sigma Aldrich Supplier No. 12669500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Potassium thiosulfate can be used:For the preparation of aqueous biphasic systems (ABS) for the extraction of organic compounds.For the synthesis of polyacrylonitrile nanoparticles by dispersion/emulsion polymerization.As a reagent for the pretreatment of graphite powder to synthesize graphene oxide from it by Hummers'sa method.As an initiator to synthesize organosilicon-based adsorbent to remove iron from crude phosphoric acid.

Specifications

CAS 10294-66-3
Molecular Formula K2O3S2
Linear Formula K2S2O3
MDL Number MFCD00036292
Quantity 500 g
Molecular Weight (g/mol) 190.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.