accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSCH3OO

CAS RN 10297-73-1

IUPAC Name: 1-(4-methanesulfonylphenyl)ethan-1-one
Synonyms: 1-(4-methanesulfonylphenyl)ethanone
Molecular Weight (g/mol): 198.24
Molecular Formula: C9H10O3S
InChi Key: KAVZYDHKJNABPC-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)S(C)(=O)=O
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)