Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[4-(methylsulfonyl)phenyl]ethan-1-one, Thermo Scientific™

Catalog No. JFD01520EB
Change view
Click to view available options
Amber Glass Bottle; 25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
JFD01520EB Amber Glass Bottle; 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JFD01520EB Supplier Thermo Scientific Chemicals Supplier No. JFD01520EB

missing translation for 'validMainframeMsg'

Specifications

CAS 10297-73-1
Assay Percent Range 97%
Packaging Amber glass bottle
Molecular Formula C9H10O3S
Linear Formula H3CC(O)C6H4SO2CH3
Quantity 25g
Synonym 4'-methylsulfonyl acetophenone, 1-4-methylsulfonyl phenyl ethanone, 4-methylsulfonylacetophenone, 4-methylsulfonyl acetophenone, 4-acetylphenyl methyl sulphone, 1-4-methylsulfonylphenyl ethanone, 4-methylsulphonylacetophenone, 1-4-methanesulfonyl-phenyl-ethanone, p-methylsulfonylacetophenone, 4-methanesulfonylacetophenone
InChI Key KAVZYDHKJNABPC-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(C=C1)S(=O)(=O)C
IUPAC Name 1-(4-methylsulfonylphenyl)ethanone
Molecular Weight (g/mol) 198.236
PubChem CID 82529
Formula Weight 198.24
Percent Purity 97%
Chemical Name or Material 1-[4-(Methylsulfonyl)phenyl]ethan-1-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.