accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10299-35-1

CH3CH3OH3COOCH3O

CAS RN 10299-35-1

IUPAC Name: 5-(acetyloxy)hexan-2-yl acetate
Synonyms: 5-(acetyloxy)hexan-2-yl acetate
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: UNAGYNUYXVBAEI-UHFFFAOYNA-N
SMILES: CC(CCC(C)OC(C)=O)OC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)