Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diacetoxyhexane 98.0+%, TCI America™
SDP

Catalog No. D34965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34965G 5 g
D349625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34965G Supplier TCI America Supplier No. D34965G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10299-35-1
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
InChI Key UNAGYNUYXVBAEI-UHFFFAOYSA-N
Synonym 2,5-Hexanediol Diacetate
PubChem CID 254550
IUPAC Name 5-acetyloxyhexan-2-yl acetate
SMILES CC(CCC(C)OC(=O)C)OC(=O)C

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 202.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Diacetoxyhexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.